2-Chloro-3,4-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A273111-100MG A273111-250MG A273111-1G
103813-130EA 51 USD
103813-130 103853-046 76875-120 76875-122 77092-840
2-Chloro-3,4-difluorobenzaldehyde 98%
2-Chloro-3,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O
MW: 176.5479 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415551
CAS Number: 1261869-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR