2-Chloro-3-(trifluoromethyl)phenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A243122-1G
A243122-25G
A243122-5G
A243122-10G
A243122-250MG
103811-506EA
8
USD
103811-506
103851-330
77090-522
77090-524
77090-526
77276-090
77276-092
2-Chloro-3-(trifluoromethyl)phenylboronic acid 97%
2-Chloro-3-(trifluoromethyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₅BClF₃O₂ MW: 224.3726 g/mol Boiling Pt: 296.5 °C (760 mm Hg) Flash Pt: 133.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD09258745 CAS Number: 957061-11-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...