2-Chloro-3-(trifluoromethyl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-α,α,α-trifluoro-m-toluidine

A129500-100G A129500-10G A129500-1G A129500-25G A129500-5G
103801-286EA 7 USD
103801-286 76971-994 76971-996 77298-344 77298-346 77313-674
2-Chloro-3-(trifluoromethyl)aniline 97%
2-Chloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MW: 195.57
Boiling Pt: 217.5 °C (760 mm Hg)
Flash Pt: 85.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04115988
CAS Number: 62476-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR