2-Chloro-1-methylpyridinium iodide 95%

Supplier: Ambeed
Warning

A100901-100G A100901-10G A100901-25G A100901-500G A100901-5G
103821-374EA 9 USD
103821-374 77107-172 77107-174 77107-176 77107-178 77282-598
2-Chloro-1-methylpyridinium iodide 95%
2-Chloro-1-methylpyridinium iodide

Precaución: for Research Use Only

Formula: C₆H₇ClIN
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00011984
CAS Number: 14338-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR