2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-imidazole

A123051-250MG A123051-1G
103800-336EA 46 USD
103800-336 77009-528 77297-920
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester 97%
2-Chloro-1-methylimidazole-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₆BClN₂O₂
Boiling Pt: 367.3±34.0 °C (760 mm Hg)
Melting Pt: 131…135 °C
Flash Pt: 175.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11100959
CAS Number: 1315281-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR