2-Chloroimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-1H-imidazole

A265274-100MG A265274-250MG A265274-1G A265274-5G
103812-716EA 27 USD
103812-716 103852-610 76977-070 76977-072 77092-280 77092-282
2-Chloroimidazole 97%
2-Chloroimidazole

Precaución: for Research Use Only

Formula: C₃H₃ClN₂
MW: 102.52 g/mol
Boiling Pt: 252.7 °C (760 mm Hg)
Melting Pt: 165…170 °C
Flash Pt: 131.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR