2-Chloro-1,3-thiazole-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A782142-1G A782142-5G A782142-250MG
103840-982EA 34 USD
103840-982 76845-236 76845-238 77268-094
2-Chloro-1,3-thiazole-5-carboxylic acid 95%
2-Chloro-1,3-thiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂ClNO₂S
Boiling Pt: 370.2 °C (760 mm Hg)
Melting Pt: 165.5…167 °C
Storage Temperature: Freezer
MDL Number: MFCD02187331
CAS Number: 101012-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR