2-Chloro-1,3-dimethylimidazolinium chloride 97% (by titrimetric analysis)

Supplier: Ambeed
Warning

Synonyms: DMC

A557547-25G A557547-5G A557547-100G A557547-10G A557547-1G
103828-724EA 5 USD
103828-724 103839-858 76843-958 76843-960 77266-318 77266-320 77307-802
2-Chloro-1,3-dimethylimidazolinium chloride 97% (by titrimetric analysis)
2-Chloro-1,3-dimethylimidazolinium chloride

Precaución: for Research Use Only

Formula: C₅H₁₀Cl₂N₂
MW: 169.05 g/mol
Melting Pt: 133…140 °C
Storage Temperature: Freezer
MDL Number: MFCD09039290
CAS Number: 37091-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR