2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate 97%

Supplier: Ambeed
Warning

Synonyms: CIP, 2-Chloro-1,3-dimethyl-4,5-dihydro-1H-imidazol-3-ium hexafluorophosphate(V)

A379172-1G A379172-100G A379172-10G A379172-25G A379172-5G
103826-642EA 7 USD
103826-642 103837-674 76840-984 77264-220 77264-222 77306-906 77306-908
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate 97%
2-Chloro-1,3-dimethylimidazolinium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₅H₁₀ClF₆N₂P
MW: 278.56 g/mol
Melting Pt: 231…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00191914
CAS Number: 101385-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR