2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
Supplier: Ambeed
|
Danger
|
A174944-100MG
A174944-250MG
A174944-1G
103806-990EA
89
USD
103806-990
103845-290
76813-342
76813-344
77082-602
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride 95%
2-Chloro-1-(pyridin-3-yl)ethanone hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₆ClNO·ClH MW: 192.043 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09033889 CAS Number: 61889-48-3 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...