3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-1-((3-fluorobenzyl)oxy)-4-nitrobenzene, 2-Chloro-1-(3-fluorobenzyloxy)-4-nitrobenzene

A102026-100G A102026-10G A102026-25G A102026-1G A102026-5G
103796-254EA 8 USD
103796-254 103821-954 77270-276 77270-278 77270-280 77308-838 77308-840
3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene 98%
3-Chloro-4-(3-fluorobenzyloxy)nitrobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉ClFNO₃
MW: 281.67 g/mol
Boiling Pt: 400.5 °C (760 mm Hg)
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD06809827
CAS Number: 443882-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR