2-Chloro-3-picoline 97%
Supplier: Ambeed
|
Warning
|
A234851-100G
A234851-10G
A234851-25G
A234851-5G
103850-742EA
5
USD
103850-742
77000-272
77000-274
77089-740
77089-742
2-Chloro-3-picoline 97%
2-Chloro-3-picoline
Precaución: for Research Use Only
Formula:
C₆H₆ClN Boiling Pt: 192.5 °C (760 mm Hg) Flash Pt: 79.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234177 CAS Number: 18368-76-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...