2-Carboxythiophene-5-boronic acid 98%

Supplier: Ambeed
Warning

A217244-250MG A217244-25G A217244-5G
103818-610EA 12 USD
103818-610 103849-342 76975-738 77303-450 77303-452
2-Carboxythiophene-5-boronic acid 98%
2-Carboxythiophene-5-boronic acid

Precaución: for Research Use Only

Formula: C₅H₅BO₄S
Melting Pt: 135…136 °C
Storage Temperature: Freezer
MDL Number: MFCD02323211
CAS Number: 465515-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR