2-Formylbenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Carboxybenzaldehyde, Phthalaldehydic acid

A281776-25G A281776-100G A281776-10G A281776-500G A281776-5G
103809-942EA 174 USD
103809-942 76837-226 77002-182 77002-184 77002-186 77002-188
2-Formylbenzoic acid 98%
2-Formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: 94…96 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00003336
CAS Number: 119-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR