2-Butylbenzo[d]isothiazol-3(2H)-one 95%

Supplier: Ambeed
Danger

A245313-100G A245313-1G A245313-25G A245313-5G
103811-634EA 7 USD
103811-634 103851-472 77261-864 77304-350 77304-352 77304-354
2-Butylbenzo[d]isothiazol-3(2H)-one 95%
2-Butylbenzo[d]isothiazol-3(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NOS
Boiling Pt: 332.921 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD09751282
CAS Number: 4299-07-4
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR