2-Butyl-5-chloroimidazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Butyl-4-chloro-5-formylimidazole, 2-(But-1-yl)-5-chloro-1H-imidazole-4-carboxaldehyde, 2-Butyl-4-chloro-1H-imidazole-5-carbaldehyde

A148416-10G A148416-500G A148416-1000G A148416-100G
103808-708EA 167 USD
103808-708 77190-694 77190-696 77317-502 77615-194
2-Butyl-5-chloroimidazole-4-carbaldehyde 97%
2-Butyl-5-chloroimidazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₁₁ClN₂O
MW: 186.6387 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01934396
CAS Number: 83857-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR