2-Bromo-3-thiophenecarboxylic acid 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromothiophene-3-carboxylic acid

A176085-1G A176085-100MG A176085-250MG
103807-116EA 23 USD
103807-116 103845-432 77082-788 77260-082 77260-084
2-Bromo-3-thiophenecarboxylic acid 96%
2-Bromo-3-thiophenecarboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₂S
Boiling Pt: 313.2 °C (760 mm Hg)
Melting Pt: 178…182 °C
Flash Pt: 143.225 °C
Storage Temperature: Ambient
MDL Number: MFCD05865213
CAS Number: 24287-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR