2-Bromo-5-thiazolecarboxylic acid 97%

Supplier: Ambeed
Warning

A138757-100G A138757-10G A138757-25G A138757-1G A138757-5G A138757-500G
103802-662EA 16 USD
103802-662 76811-668 76811-670 77299-020 77315-714 77315-716 77670-988
2-Bromo-5-thiazolecarboxylic acid 97%
2-Bromo-5-thiazolecarboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrNO₂S
Melting Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD04115730
CAS Number: 54045-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR