2-Bromothiazole-5-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A202579-100MG
A202579-10G
A202579-250MG
A202579-5G
A202579-1G
103817-402EA
47
USD
103817-402
103848-046
76785-396
76785-398
76785-400
77086-160
77302-816
2-Bromothiazole-5-carbonitrile 98%
2-Bromothiazole-5-carbonitrile
Precaución: for Research Use Only
Formula:
C₄HBrN₂S Boiling Pt: 276.1 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09909651 CAS Number: 440100-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...