2-Bromostyrene 97% stabilized
Supplier: Ambeed
|
Warning
|
Stabilization:
Stabilised with 4-tert-butylpyrocatechol (TBC)
A299743-1G
A299743-10G
103814-242EA
15
USD
103814-242
103835-392
76838-082
77550-814
2-Bromostyrene 97% stabilized
2-Bromostyrene
Precaución: for Research Use Only
Formula:
C₈H₇Br MW: 183.05 g/mol Boiling Pt: 209.9 °C (760 mm Hg) Melting Pt: 7 °C Flash Pt: 86 °C Storage Temperature: Freezer |
MDL Number:
MFCD00000076 CAS Number: 2039-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...