2-Bromoquinoxaline 98%

Supplier: Ambeed
Warning

A104314-10G A104314-25G A104314-1G A104314-5G A104314-250MG
103796-688EA 18 USD
103796-688 103822-398 77093-634 77093-636 77283-200 77283-202 77684-948
2-Bromoquinoxaline 98%
2-Bromoquinoxaline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
MW: 209.04 g/mol
Melting Pt: 56…60 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD07368206
CAS Number: 36856-91-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR