2-Bromoquinoline-4-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A201707-1G
A201707-5G
A201707-250MG
103817-320EA
131
USD
103817-320
103847-960
77086-036
77086-038
77260-802
2-Bromoquinoline-4-carboxylic acid 97%
2-Bromoquinoline-4-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₆BrNO₂ MW: 252.0600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08705669 CAS Number: 15733-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...