2-Bromoquinoline 97%

Supplier: Ambeed
Warning

A168644-1G A168644-5G A168644-100G A168644-10G A168644-500G A168644-25G A168644-250MG
103806-308EA 6 USD
103806-308 103844-562 77081-618 77081-620 77259-848 77259-850 77259-852 77300-936 77746-129
2-Bromoquinoline 97%
2-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 292.5 °C (760 mm Hg)
Melting Pt: 48…49 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00234480
CAS Number: 2005-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR