2-Bromoquinolin-8-ol 95%

Supplier: Ambeed
Warning

A116240-100MG A116240-10G A116240-1G A116240-5G A116240-250MG
103798-698EA 88 USD
103798-698 103824-560 77009-192 77009-194 77297-438 77297-440 77889-798
2-Bromoquinolin-8-ol 95%
2-Bromoquinolin-8-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 367.0±22.0 °C (760 mm Hg)
Melting Pt: 81…82 °C
Flash Pt: 175.8 °C
Storage Temperature: Ambient
MDL Number: MFCD18413576
CAS Number: 139399-61-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR