2-Bromopyrimidine-4-carbonitrile 98%

Supplier: Ambeed
Warning

A262455-250MG A262455-1G
103812-566EA 200 USD
103812-566 103852-462 76816-034 77092-082
2-Bromopyrimidine-4-carbonitrile 98%
2-Bromopyrimidine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂BrN₃
MW: 183.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13193321
CAS Number: 1209459-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR