2-Bromopyrimidine 98%
Supplier: Ambeed
|
Warning
|
A354927-100G
A354927-10G
A354927-500G
A354927-5G
103837-140EA
15
USD
103837-140
77033-136
77033-138
77033-140
77306-672
2-Bromopyrimidine 98%
2-Bromopyrimidine
Precaución: for Research Use Only
Formula:
C₄H₃BrN₂ Boiling Pt: 249.9 °C (760 mm Hg) Melting Pt: 55…57 °C Storage Temperature: Freezer |
MDL Number:
MFCD00014601 CAS Number: 4595-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...