(2-Bromo-4-pyridinyl)methanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopyridine-4-methanol, 2-Bromo-4-pyridinemethanol

A186800-1G A186800-5G A186800-10G
103815-964EA 10 USD
103815-964 103846-552 77084-238 77084-240 77111-604
(2-Bromo-4-pyridinyl)methanol 98%
(2-Bromo-4-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Melting Pt: 60…65 °C
Flash Pt: 145.3 °C
Storage Temperature: Ambient
MDL Number: MFCD04039313
CAS Number: 118289-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR