2-Bromo-4-formylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-4-pyridinecarboxaldehyde, 2-Bromopyridine-4-carboxaldehyde
A148691-100G
A148691-250MG
A148691-1G
A148691-25G
A148691-5G
103803-956EA
19
USD
103803-956
103842-074
77095-742
77095-744
77317-536
77317-538
77317-540
2-Bromo-4-formylpyridine 97%
2-Bromo-4-formylpyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrNO MW: 186.01 g/mol Flash Pt: 118.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04039311 CAS Number: 118289-17-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...