1-Bromo-2-ethynylbenzene 97%
Supplier: Ambeed
|
Warning
|
A476960-1G
A476960-25G
A476960-5G
A476960-100G
A476960-10G
103828-090EA
16
USD
103828-090
103839-160
76843-094
76843-096
76843-098
77196-730
77196-732
1-Bromo-2-ethynylbenzene 97%
1-Bromo-2-ethynylbenzene
Precaución: for Research Use Only
Formula:
C₈H₅Br Boiling Pt: 92…93 °C (20 mm Hg) Flash Pt: 63.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863358 CAS Number: 766-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...