2-Bromooxazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A136738-1G A136738-250MG
103802-342EA 61 USD
103802-342 77298-864 77592-590
2-Bromooxazole-4-carboxylic acid 95%
2-Bromooxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₂BrNO₃
MW: 191.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845654
CAS Number: 1167055-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR