2-Bromonicotinonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-3-cyanopyridine
A230607-100G
A230607-10G
A230607-250MG
A230607-1G
A230607-5G
A230607-25G
103819-558EA
12
USD
103819-558
103850-458
76976-138
76976-140
76976-142
77089-322
77089-324
77303-880
2-Bromonicotinonitrile 98%
2-Bromonicotinonitrile
Precaución: for Research Use Only
Formula:
C₆H₃BrN₂ MW: 183.01 g/mol Boiling Pt: 296.9 °C (760 mm Hg) Flash Pt: 133.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00234140 CAS Number: 20577-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...