2-Bromo-3-formylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromopyridine-3-carboxaldehyde, 2-Bromonicotinaldehyde, 2-Bromo-3-pyridinecarboxaldehyde

A207032-100G A207032-10G A207032-500G A207032-25G A207032-5G A207032-1G
103817-756EA 9 USD
103817-756 103848-436 77012-830 77012-832 77012-834 77086-702 77086-704 77303-020
2-Bromo-3-formylpyridine 98%
2-Bromo-3-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 70…74 °C
Flash Pt: 115.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04966945
CAS Number: 128071-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR