2-Bromonaphthalen-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-naphthalenol

A233669-1G A233669-25G A233669-5G A233669-250MG
103810-902EA 14 USD
103810-902 103850-660 77089-624 77089-626 77089-628 77261-592
2-Bromonaphthalen-1-ol 97%
2-Bromonaphthalen-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Boiling Pt: 313.7 °C (760 mm Hg)
Melting Pt: 44…46 °C
Flash Pt: 143.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00274158
CAS Number: 771-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR