2-Bromo-N,N-dimethylpyridin-4-amine 95%

Supplier: Ambeed
Warning

A228102-250MG A228102-5G A228102-1G A228102-10G
103819-360EA 43 USD
103819-360 103850-268 76976-020 77089-062 77303-816 77881-582
2-Bromo-N,N-dimethylpyridin-4-amine 95%
2-Bromo-N,N-dimethylpyridin-4-amine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
Flash Pt: 126.608 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11036210
CAS Number: 396092-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR