2-Bromo-N,N-dimethylethanamine hydrobromide 97%

Supplier: Ambeed
Warning

A271012-100G A271012-10G A271012-25G A271012-5G A271012-1G A271012-500G
103813-026EA 9 USD
103813-026 103852-936 76692-842 76692-844 77092-708 77092-710 77305-064 77626-410
2-Bromo-N,N-dimethylethanamine hydrobromide 97%
2-Bromo-N,N-dimethylethanamine hydrobromide

Precaución: for Research Use Only

Formula: C₄H₁₁Br₂N
Storage Temperature: Refrigerator
MDL Number: MFCD00040375
CAS Number: 2862-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR