2-Bromo-N,N-dimethylaniline 97%

Supplier: Ambeed
Warning

A160266-100G A160266-10G A160266-1G A160266-25G A160266-5G
103805-354EA 5 USD
103805-354 103843-584 77259-592 77259-594 77300-456 77319-502 77319-504
2-Bromo-N,N-dimethylaniline 97%
2-Bromo-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 225.9 °C (760 mm Hg)
Flash Pt: 90.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00013522
CAS Number: 698-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR