2-(Bromomethyl)-1,3-dioxolane 97%

Supplier: Ambeed
Warning

A346317-1000G A346317-500G A346317-25G A346317-5G
103836-922EA 5 USD
103836-922 76648-432 76648-434 76648-436 77306-572 77306-574
2-(Bromomethyl)-1,3-dioxolane 97%
2-(Bromomethyl)-1,3-dioxolane

Precaución: for Research Use Only

Formula: C₄H₇BrO₂
Boiling Pt: 178.7 °C (760 mm Hg)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003214
CAS Number: 4360-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR