2-Bromoisonicotinonitrile 95%

Supplier: Ambeed
Warning

A137674-100G A137674-10G A137674-1G A137674-25G A137674-5G
103802-482EA 5 USD
103802-482 77027-358 77027-360 77315-456 77315-458 77315-460
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 243 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD07367879
CAS Number: 10386-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR