2-Bromoisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid

A139877-100G A139877-10G A139877-500G A139877-25G A139877-5G
77285-692EA 80.34 USD
77285-692 77285-694 77285-696 77299-094 77299-096
2-Bromoisonicotinic acid 98%
2-Bromoisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Boiling Pt: 447.2 °C (760 mm Hg)
Melting Pt: 229…231 °C
Flash Pt: 224.3 °C
Storage Temperature: Ambient
MDL Number: MFCD01646069
CAS Number: 66572-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR