2-Bromoimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A455866-100MG A455866-250MG A455866-50MG A455866-1G A455866-25G A455866-5G A455866-10G
103827-850EA 22 USD
103827-850 103838-924 76789-416 76789-418 76789-420 76842-764 76842-766 76842-768 77669-046
2-Bromoimidazo[1,2-a]pyridine 97%
2-Bromoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09834778
CAS Number: 112581-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR