2-Bromofluorene 97%

Supplier: Ambeed
Warning

A430508-1000G A430508-25G A430508-5G A430508-100G A430508-10G A430508-500G
103810-334EA 225 USD
103810-334 103838-576 76842-260 76842-262 76842-264 77102-254 77102-256 77102-258
2-Bromofluorene 97%
2-Bromofluorene

Precaución: for Research Use Only

Formula: C₁₃H₉Br
Boiling Pt: 185 °C (760 mm Hg)
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00001115
CAS Number: 1133-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR