2-Bromoethyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

Synonyms: 2-Bromoethyl triflate

A922344-100MG A922344-250MG A922344-1G
103841-410EA 320 USD
103841-410 76820-966 76820-968 77308-520
2-Bromoethyl trifluoromethanesulfonate 95%
2-Bromoethyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₃H₄BrF₃O₃S
MW: 257.026 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656439
CAS Number: 103935-47-3
UN: 2927
ADR: 6.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR