2-Bromobenzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromophenyl)methanol

A335482-1000G A335482-100G A335482-10G A335482-500G A335482-25G A335482-5G
103810-158EA 314 USD
103810-158 103836-662 77106-564 77106-566 77106-568 77106-570 77306-488 77306-490
2-Bromobenzyl alcohol 98%
2-Bromobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₇BrO
Boiling Pt: 261.8 °C (760 mm Hg)
Melting Pt: 78…80 °C
Flash Pt: 112.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00004600
CAS Number: 18982-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR