2-Bromobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: o-Bromobenzoic Acid

A239277-100G A239277-500G A239277-5G A239277-25G
103809-696EA 73 USD
103809-696 76874-384 76874-386 76874-388 77304-150
2-Bromobenzoic acid 98%
2-Bromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 147…150 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR