2-Bromobenzo[b]furan 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-1-benzofuran, 2-Bromobenzofuran

A105217-100MG A105217-250MG
103796-856EA 33 USD
103796-856 103822-584 77257-414 77745-796
2-Bromobenzo[b]furan 95%
2-Bromobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₅BrO
MW: 197.029 g/mol
Boiling Pt: 233.8 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Freezer
MDL Number: MFCD02681987
CAS Number: 54008-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR