2-Bromobenzo[b]thiophene 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-benzo[b]thiophene

A171037-10G A171037-25G A171037-5G
103806-580EA 13 USD
103806-580 103844-854 77259-926 77301-086 77301-088
2-Bromobenzo[b]thiophene 96%
2-Bromobenzo[b]thiophene

Precaución: for Research Use Only

Formula: C₈H₅BrS
MW: 213.0943 g/mol
Boiling Pt: 284.7 °C (760 mm Hg)
Melting Pt: 37…43 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08435846
CAS Number: 5394-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR