2-Bromo-9,10-anthraquinone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromoanthracene-9,10-dione, 2-Bromo-9,10-anthracenedione

A212808-25G A212808-100G A212808-10G A212808-500G A212808-1G A212808-5G
103818-236EA 9 USD
103818-236 103848-932 77087-406 77112-482 77112-484 77112-486 77303-258 77303-260
2-Bromo-9,10-anthraquinone 98%
2-Bromo-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₇BrO₂
MW: 287.11 g/mol
Boiling Pt: 443.4 °C (760 mm Hg)
Melting Pt: 208 °C
Flash Pt: 160.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00474541
CAS Number: 572-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR