2-Bromoanthracene 97%

Supplier: Ambeed
Warning

A107649-100G A107649-10G A107649-250MG A107649-25G A107649-1G A107649-5G
103797-226EA 13 USD
103797-226 103822-992 77107-772 77107-774 77107-776 77107-778 77283-632 77309-850
2-Bromoanthracene 97%
2-Bromoanthracene

Precaución: for Research Use Only

Formula: C₁₄H₉Br
MW: 257.1300 g/mol
Boiling Pt: 389.7 °C (760 mm Hg)
Melting Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD07784002
CAS Number: 7321-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR