2'-Bromoacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: o-Bromoacetophenone

A238597-25G A238597-5G A238597-1000G A238597-100G A238597-10G A238597-1G A238597-500G
103851-022EA 10 USD
103851-022 77090-144 77090-146 77098-418 77098-420 77098-422 77098-424 77098-426
2'-Bromoacetophenone 98%
2'-Bromoacetophenone

Precaución: for Research Use Only

Formula: C₈H₇BrO
MW: 199.05 g/mol
Boiling Pt: 249 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000067
CAS Number: 2142-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR