2-Bromo-9-fluorenone 97%

Supplier: Ambeed
Warning

A100796-100G A100796-10G A100796-1G A100796-25G A100796-500G A100796-5G
103821-326EA 6 USD
103821-326 76994-472 76994-474 76994-476 76994-478 76994-480 77282-562
2-Bromo-9-fluorenone 97%
2-Bromo-9-fluorenone

Precaución: for Research Use Only

Formula: C₁₃H₇BrO
Boiling Pt: 392.8 °C (760 mm Hg)
Melting Pt: 143…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00010789
CAS Number: 3096-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR